Learn More
DL-alpha-Amino-2-thiopheneacetic acid methyl ester hydrochloride, 98%
Product Code. 11410658
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 11410658
Brand: Thermo Scientific Alfa Aesar B25190.03
Request Bulk or Custom Format
CAS: 132289-66-8 | C7H10ClNO2S | 207.67 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 132289-66-8
Molecular Formula C7H10ClNO2S
Molecular Weight (g/mol) 207.67
MDL Number MFCD04039779
InChI Key NZEQIXANVTXFEP-UHFFFAOYNA-N
Synonym methyl 2-amino-2-thiophen-2-yl acetate hydrochloride, dl-alpha-aminothiophene-2-acetic acid methyl ester hydrochloride
PubChem CID 24212017
SMILES Cl.COC(=O)C(N)C1=CC=CS1
Specifications
Melting Point ∼180°C (decomposition)
Quantity 1 g
Sensitivity Hygroscopic
IUPAC Name methyl 2-amino-2-(thiophen-2-yl)acetate hydrochloride
Formula Weight 207.67
Percent Purity 98%
Chemical Name or Material DL-alpha-Amino-2-thiopheneacetic acid methyl ester hydrochloride