missing translation for 'onlineSavingsMsg'
Learn More

DL-Arginine hydrochloride, 98%, anhydrous, Thermo Scientific Chemicals

Catalog Number p-4323539
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
32042-43-6
C6H14N4O2·ClH
210.67
KWTQSFXGGICVPE-UHFFFAOYSA-N
dl-arginine hydrochloride, 2-amino-5-guanidinopentanoic acid hydrochloride, argininhydrochlorid, dl-arginine hcl, h-dl-arg-oh.xhcl, dl-+/--arginine hydrochloride, 1-hydrochloride arginine, arginine monochloride, h-dl-arg-oh.hcl, wln: muyzm3yzvq &gh
85880
2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
C(CC(C(=O)O)N)CN=C(N)N.Cl
This item is not returnable. View return policy
32042-43-6
C6H14N4O2·ClH
210.67
KWTQSFXGGICVPE-UHFFFAOYSA-N
dl-arginine hydrochloride, 2-amino-5-guanidinopentanoic acid hydrochloride, argininhydrochlorid, dl-arginine hcl, h-dl-arg-oh.xhcl, dl-+/--arginine hydrochloride, 1-hydrochloride arginine, arginine monochloride, h-dl-arg-oh.hcl, wln: muyzm3yzvq &gh
85880
2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
C(CC(C(=O)O)N)CN=C(N)N.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 200.0°C to 203.0°C
Color White
Loss on Drying 3% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5%
Packaging Glass Bottle
Linear Formula H2NC(=NH)NH(CH2)3CH(NH2)CO2H·HCl
Quantity 1 g
Solubility Information Solubility in water: soluble.
Formula Weight 210.67
Percent Purity 98%
Physical Form Powder
Chemical Name or Material DL-Arginine hydrochloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'