Learn More
DL-Isocitric Acid Lactone, TRC
Product Code. 30134777
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134777 500 mg 500mg
30003995 100 mg 100mg
30008172 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134777 Supplier Toronto Research Chemicals Supplier No. TRCI627103500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4702-32-3
Molecular Formula C6 H6 O6
Molecular Weight (g/mol) 174.11
Synonym 1,4-Lactone-3-carboxy-2,3-dideoxypentaric acid, Isocitric lactone, Tetrahydro-5-oxo-2,3-furandicarboxylic acid
IUPAC Name 5-oxotetrahydrofuran-2,3-dicarboxylic acid
SMILES OC(=O)C1CC(=O)OC1C(=O)O
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C6H6O6/c7-3-1-2(5(8)9)4(12-3)6(10)11/h2,4H,1H2,(H,8,9)(H,10,11)
Formula Weight 174.0164
Chemical Name or Material DL-Isocitric Acid Lactone