Learn More
DL-Leucine-d7 (iso-propyl-d7), CDN
Specifications
Specifications
| Chemical Name or Material | DL-Leucine-4,5,5,5,5',5',5'-D7 |
| CAS | 259225-40-6 |
| Molecular Formula | C6 D7 H6 N O2 |
| Quantity | 0.1 g |
| Synonym | Norvaline-4,5,5,5-d4, 4-(methyl-d3)- (9 CI), DL-Leucine-4,5,5,5,5',5',5'-d7, DL-Leucine-d7 (iso-propyl-d7), DL-Leucine-d7, 2-Amino-(4,5,5,5-tetradeuterio)-4-(trideuteriomethyl)pentanoic acid, DL-Leucine-4,5,5,5,5',5',5'-D7, DL-Leucine-4,5,5,5,5',5',5'-D7 |
| InChI Formula | InChI=1 S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5 H,3,7H2,1-2H3,(H,8,9)/i1D3,2D3,4 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(CC(N)C(=O)O)C([2 H])([2 H])[2 H] |
| IUPAC Name | 2-amino-4,5,5,5-tetradeuterio-4-(trideuteriomethyl)pentanoic acid |
| Molecular Weight (g/mol) | 138.2161 |
| Formula Weight | 138.1386 g/mol |
| Show More |