Learn More
DL-Xylose, MedChemExpress
Product Code. 16448586
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16448586 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16448586 Supplier Medchem Express Supplier No. HYB1070
MedChemExpress DL-Xylose is an intermediate of organic synthesis.
DL-Xylose is an intermediate of organic synthesis.
Specifications
Chemical Name or Material DL-Xylose
CAS 25990-60-7
Color White
Purity Grade Notes Research
Molecular Formula C5H10O5
Quantity 100 mg
Synonym (±)-Xylos
Solubility Information H2O : 125 mg/mL (832.61 mM; Need ultrasonic)
SMILES O=C[C@@H]([C@H]([C@@H](CO)O)O)O
Molecular Weight (g/mol) 150.13
Formula Weight 150.13
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only