missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLC1/DLEC1 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06065PE
This item is not returnable.
View return policy
Description
DLC1/DLEC1 Polyclonal antibody specifically detects DLC1/DLEC1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| DLC1/DLEC1 | |
| PBS | |
| Antigen and protein A Affinity-purified | |
| Purified |
| Polyclonal | |
| Produced in rabbits immunized with E. coli-derived Human DLC1/DLEC1 fragment. | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction