Learn More
DLD Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI8A10, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF503388
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes the L protein of the mitochondrial glycine cleavage system. The L protein, also named dihydrolipoamide dehydrogenase, is also a component of the pyruvate dehydrogenase complex, the alpha-ketoglutarate dehydrogenase complex, and the branched-chain alpha-keto acide dehydrogenase complex. Mutations in this gene have been identified in patients with E3-deficient maple syrup urine disease and lipoamide dehydrogenase deficiency.Specifications
| DLD | |
| Monoclonal | |
| Unconjugated | |
| DLD | |
| DLDD, DLDH, E3, GCSL, LAD, PHE3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 13382, 1738, 298942, 403978 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Western Blot | |
| OTI8A10 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O08749, P09622, P49819, Q6P6R2 | |
| Dld | |
| Full length human recombit protein of human DLD produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.