missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ DLEC/CLEC4C/BDCA-2 Antibody (CLEC4C/3400) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26924AF532
This item is not returnable.
View return policy
Description
DLEC/CLEC4C/BDCA-2 Monoclonal antibody specifically detects DLEC/CLEC4C/BDCA-2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| DLEC/CLEC4C/BDCA-2 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| BDCA2, BDCA-2, BDCA2MGC125791, Blood dendritic cell antigen 2, blood dendritic cell antigen 2 protein, CD303, CD303 antigen, CLECSF11, CLECSF11PRO34150, CLECSF7, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 11, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 7, C-type lectin domain family 4 member C, C-type lectin superfamily member 7, dendritic cell lectin b, Dendritic lectin, DLEC, DLECMGC125792, HECL, HECLMGC125793, MGC125789, PRO34150 | |
| Recombinant fragment (around aa1-250) of human DLEC/CLEC4C/BDCA-2 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Dendritic Cell Markers | |
| 170482 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| CLEC4C/3400 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction