missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ DLEC/CLEC4C/BDCA-2 Antibody (CLEC4C/3400) [Alexa Fluor™ 750], Novus Biologicals™

Product Code. 30061226 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30061226 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30061226 Supplier Novus Biologicals™ Supplier No. NBP326924AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DLEC/CLEC4C/BDCA-2 Monoclonal antibody specifically detects DLEC/CLEC4C/BDCA-2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen DLEC/CLEC4C/BDCA-2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CLEC4C/3400
Conjugate Alexa Fluor 750
Formulation 50 mM Sodium Borate
Gene Alias BDCA2, BDCA-2, BDCA2MGC125791, Blood dendritic cell antigen 2, blood dendritic cell antigen 2 protein, CD303, CD303 antigen, CLECSF11, CLECSF11PRO34150, CLECSF7, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 11, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 7, C-type lectin domain family 4 member C, C-type lectin superfamily member 7, dendritic cell lectin b, Dendritic lectin, DLEC, DLECMGC125792, HECL, HECLMGC125793, MGC125789, PRO34150
Host Species Mouse
Immunogen Recombinant fragment (around aa1-250) of human DLEC/CLEC4C/BDCA-2 protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Dendritic Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 170482
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.