missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SPATA5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29450
This item is not returnable.
View return policy
Description
SPATA5 Polyclonal antibody specifically detects SPATA5 in Human samples. It is validated for Immunoprecipitation
Specifications
| SPATA5 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 166378 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation | |
| Unconjugated | |
| AFG2ATPase family gene 2 homolog, ATPase family protein 2 homolog, SPAFspermatogenesis associated factor SPAF, spermatogenesis associated 5, Spermatogenesis-associated factor protein, spermatogenesis-associated protein 5 | |
| Between 843 and 893 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction