Learn More
DLEC/CLEC4C/BDCA-2 Antibody (LFB patent anti-BDCA-2) [Janelia Fluor™ 669], Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-28633JF669
Product Code. 30172031
  • € 431.00 / 0.10mL
Description
DLEC/CLEC4C/BDCA-2 Monoclonal antibody specifically detects DLEC/CLEC4C/BDCA-2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DLEC/CLEC4C/BDCA-2
Monoclonal
Janelia Fluor 669
BDCA2, BDCA-2, BDCA2MGC125791, Blood dendritic cell antigen 2, blood dendritic cell antigen 2 protein, CD303, CD303 antigen, CLECSF11, CLECSF11PRO34150, CLECSF7, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 11, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 7, C-type lectin domain family 4 member C, C-type lectin superfamily member 7, dendritic cell lectin b, Dendritic lectin, DLEC, DLECMGC125792, HECL, HECLMGC125793, MGC125789, PRO34150
CLEC4C
0.1 mL
Dendritic Cell Markers
170482
Store at 4°C in the dark.
IgG
Flow Cytometry, ELISA, Functional Assay
LFB patent anti-BDCA-2
50mM Sodium Borate
Human
Protein A purified