missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dlg4 Mouse anti-Rat, HRP, Clone:6G6, Abnova™
Mouse monoclonal antibody raised against recombinant rat Dlg4.
Brand: Abnova MAB17317.100ug
This item is not returnable.
View return policy
Specifications
| Dlg4 | |
| Monoclonal | |
| HRP | |
| Antibody Microarray Immunocytochemistry (1:100) Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000) Western Blot (1:250) The optimal working dilution should be determined by the end user. |
|
| Dlg4 | |
| Mouse | |
| 100 μg | |
| Rat | |
| Antibody | |
| IgG2a |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 6G6 | |
| discs, large homolog 4 (Drosophila) | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
| Dlgh4, PSD95, Sap90 | |
| Recombinant protein corresponding to rat Dlg4. | |
| 29495 | |
| Store at -20°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction