missing translation for 'onlineSavingsMsg'
Learn More
Learn More
15-PGDH/HPGD Antibody (OTI2C10), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70933MFV450
This item is not returnable.
View return policy
Description
15-PGDH/HPGD Monoclonal antibody specifically detects 15-PGDH/HPGD in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| 15-PGDH/HPGD | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| OTI2C10 | |
| Flow Cytometry | |
| 15-hydroxyprostaglandin dehydrogenase [NAD+], 15-PGDH, EC 1.1.1, EC 1.1.1.141, hydroxyprostaglandin dehydrogenase 15-(NAD), NAD+-dependent 15-hydroxyprostaglandin dehydrogenase, PGDH1PGDH, Prostaglandin dehydrogenase 1, SDR36C1, short chain dehydrogenase/reductase family 36C, member 1 | |
| Full length human recombinant protein of human HPGD (NP_000851) produced in HEK293T cell. | |
| 0.1 mL | |
| Cancer, Immunology, Innate Immunity | |
| 3248 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction