missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DLG7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576093
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
DLG7 is a potential cell cycle regulator that may play a role in carcinogenesis of cancer cells and mitotic phosphoprotein regulated by the ubiquitin-proteasome pathway. The protein is also a key regulator of adherens junction integrity and differentiation that may be involved in CDH1-mediated adhesion and signaling in epithelial cells.
Specifications
| DLG7 | |
| Polyclonal | |
| Unconjugated | |
| DLGAP5 | |
| C77459; C86398; DAP-5; discs large homolog 7; discs large homolog associated protein 5; discs, large (Drosophila) homolog-associated protein 5; discs, large homolog 7; discs, large homolog-associated protein 5; disks large-associated protein 5; Disks large-associated protein DLG7; DLG associated protein 5; DLG7; DLGAP5; Hepatoma up-regulated protein; hepatoma up-regulated protein homolog; HURP; Kiaa0008; mKIAA0008 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9787 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15398 | |
| DLGAP5 | |
| Recombinant full length Human DLG7. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering