missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLGAP4 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98641DL594
This item is not returnable.
View return policy
Description
DLGAP4 Polyclonal antibody specifically detects DLGAP4 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| DLGAP4 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 594 | |
| DAP4FLJ20588, discs, large (Drosophila) homolog-associated protein 4, disks large associated protein 4, disks large-associated protein 4, DLP4, KIAA0964MGC131862, PSD-95/SAP90-binding protein 4, RP5-977B1.6, SAP90/PSD-95-associated protein 4, SAPAP-4, SAPAP4DAP-4 | |
| Produced in rabbits immunized with E. coli-derived Human DLGAP4 fragment. | |
| 0.1 mL | |
| Signal Transduction | |
| 22839 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction