missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLL1 Antibody (MM0247- 2X38), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12266AF488
This item is not returnable.
View return policy
Description
DLL1 Monoclonal specifically detects DLL1 in Human samples. It is validated for Western Blot.
Specifications
| DLL1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| O00548 | |
| DLL1 | |
| Human recombinant DLL1 | |
| 0.1 mL | |
| Neuroscience, Stem Cell Signaling Pathway | |
| 28514 | |
| Store at 4C in the dark. | |
| IgG2 |
| Western Blot | |
| MM0247- 2X38 | |
| Western Blot | |
| Delta, delta (Drosophila)-like 1, Delta1, delta-like 1 (Drosophila), delta-like protein 1, DL1, Drosophila Delta homolog 1, H-Delta-1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction