missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLL1 Antibody (MM0247- 2X38), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12266AF532
This item is not returnable.
View return policy
Description
DLL1 Monoclonal specifically detects DLL1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| DLL1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| DLL1 | |
| Human recombinant DLL1 | |
| Primary | |
| Human | |
| IgG2 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0247- 2X38 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Delta, delta (Drosophila)-like 1, Delta1, delta-like 1 (Drosophila), delta-like protein 1, DL1, Drosophila Delta homolog 1, H-Delta-1 | |
| Mouse | |
| 0.1 mL | |
| 28514 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction