Learn More
Invitrogen™ DLL1 Polyclonal Antibody
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  Invitrogen™ PA519106
Product Code. 11347083
  • € 522.00 / 100µg
Description
This antibody is predicted to react with human based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
Delta-like protein 1 (DLL1) is a transmembrane protein characterized by epidermal growth factor (EGF)-like repeats. It is a ligand for Notch receptor. It blocks the differentiation of progenitor cells into the B-cell lineage while promoting the emergence of a population of cells with the characteristics of a T-cell/NK-cell precursor.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DLL1
Polyclonal
Unconjugated
DLL1
C-Delta-1; Delta; delta like canonical Notch ligand 1; delta like-1; DELTA1; delta-like 1; delta-like 1 (Drosophila); delta-like protein 1; DL1; Dll1; Dll1-derived cell-associated form; Dll1-EC; Dll1-IC; Dll1-intracellular form; Dll1-soluble form; Dll1-TMIC; Drosophila Delta homolog 1; H-Delta-1; LOW QUALITY PROTEIN: delta-like protein 1; OTTHUMP00000017690; UNQ146/PRO172
Goat
Ammonium Sulfate Precipitation
RUO
28514, 84010
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
O00548, P97677
DLL1
Synthetic peptide sequence (ATQRHLTVGEEWSQD) corresponding to the internal amino acids of DLL1 (aa 148-162).
100 μg
Primary
Human, Rat