Learn More
Novus Biologicals™ DLL3 Antibody (Dragonfly patent anti-DLL3) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28318-100ug
 View more versions of this product
Product Code. 30044721
  • € 338.00 / 100µg
Description
DLL3 Monoclonal antibody specifically detects DLL3 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DLL3
Monoclonal
Unconjugated
Q9NYJ7
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
Dragonfly patent anti-DLL3
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
delta (Drosophila)-like 3, Delta3, delta-like 3 (Drosophila), delta-like protein 3, Drosophila Delta homolog 3, SCDO1delta3
DLL3
100 μg
Cell Biology, Cell Cycle and Replication, Neuroscience
10683