missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DLL3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA526336

Product Code. 17166029

  • € 568.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

DLL3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Delta-like protein 3 (DLL3) is a ligand for the Notch signaling pathway. It inhibits primary neurogenesis. DLL3 plays a role in the formation of somite boundaries during segmentation of the paraxial mesoderm. Defects in DLL3 are the cause of Spondylocostal dysostosis autosomal recessive type 1 (SCDO1). Mutations in DLL3 gene cause truncal shortening relative to their limbs, which leads to abdominal protrusion, abnormal spinal curvature and sometimes a plagiocephaly-torticollis sequence. It may be required to divert neurons along a specific differentiation pathway.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DLL3
Polyclonal
Unconjugated
Dll3
delta like canonical Notch ligand 3; Delta3; delta-like 3; delta-like 3 (Drosophila); delta-like protein 3; Dll3; Drosophila Delta homolog 3; M-Delta-3; pu; pudgy; SCDO1
Rabbit
Antigen affinity chromatography
RUO
10683, 114125, 13389
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.48 mg/mL
PBS with 0.09% sodium azide; pH 7.4
O88516, O88671, Q9NYJ7
Dll3
KLH conjugated synthetic peptide between 519-548 amino acids from the C-terminal region of human DLL3.
200 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.