missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLL4 Antibody (447506) [DyLight 488], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB1506K
This item is not returnable.
View return policy
Description
DLL4 Monoclonal antibody specifically detects DLL4 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| DLL4 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 447506 | |
| Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| delta 4, Delta 4 precursor, delta ligand 4, delta4, delta-like 4 (Drosophila), delta-like 4 homolog, delta-like 4 homolog (Drosophila), delta-like 4 protein, delta-like protein 4, Drosophila Delta homolog 4, hdelta2, MGC126344, notch ligand delta-2, notch ligand DLL4 | |
| Mouse myeloma cell line NS0-derived recombinant human DLL4, Ser27-Pro524, Accession # Q9NR61 | |
| 0.1 mL | |
| Neuroscience | |
| 54567 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction