missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DLL4 (delta-like 4) Monoclonal Antibody (HMD4-2)

Hamster Monoclonal Antibody
Brand: Invitrogen™ MA528213
This item is not returnable.
View return policy
Description
Hamster anti Mouse Delta-Like Protein 4 antibody, clone HMD4-2 recognizes mouse Delta-like protein 4 (DLL4), one of the five major ligands of the Notch signalling pathway, which is activated through the binding of specific ligands to the Notch receptors Notch 1-4.
Delta-like ligand 4 (DLL4), also know as Delta-4, is a homologue of the Drosophila delta protein, and functions as a transmembrane bound ligand to the notch receptor Notch1. DLL4 is expressed in the vasculature and plays a critical role in vascular development. It is induced by vascular endothelial growth factor (VEGF), as a negative feedback regulator to regulate angiogenic sprouting and promote the formulation of a differentiated vascular network. DLL4 has been found to be strongly expressed in tumor vessels of clear-cell renal tumors and bladder cancer, and inhibition of DLL4 results in increased vascular proliferation but defective maturation. This in turn leads to a decrease in tumor growth, with no apparent toxicity.
Specifications
| DLL4 (delta-like 4) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| Q9JI71 | |
| Dll4 | |
| Recombinant mouse DLL4. | |
| 250 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
| HMD4-2 | |
| Unconjugated | |
| Dll4 | |
| AOS6; delta 4; delta ligand 4; delta like canonical Notch ligand 4; Delta4; delta-like 4 (Drosophila); delta-like 4 homolog; delta-like 4 protein; delta-like protein 4; DLL4; Drosophila Delta homolog 4; hdelta2; MGC126344; notch ligand delta-2; notch ligand DLL4; UNQ1895/PRO4341 | |
| Hamster | |
| Protein A | |
| RUO | |
| 54485 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction