missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DLL4 Recombinant Rabbit Monoclonal Antibody (003), HRP
GREENER_CHOICE

Product Code. 30019360
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30019360 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30019360 Supplier Invitrogen™ Supplier No. MA552153

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

DLL4 Recombinant Monoclonal Antibody for ELISA

Delta-like ligand 4 (DLL4), also know as Delta-4, is a homologue of the Drosophila delta protein, and functions as a transmembrane bound ligand to the notch receptor Notch1. DLL4 is expressed in the vasculature and plays a critical role in vascular development. It is induced by vascular endothelial growth factor (VEGF), as a negative feedback regulator to regulate angiogenic sprouting and promote the formulation of a differentiated vascular network. DLL4 has been found to be strongly expressed in tumor vessels of clear-cell renal tumors and bladder cancer, and inhibition of DLL4 results in increased vascular proliferation but defective maturation. This in turn leads to a decrease in tumor growth, with no apparent toxicity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DLL4
Applications ELISA
Classification Recombinant Monoclonal
Clone 003
Concentration 100 μg/mL
Conjugate HRP
Formulation PBS with no preservative
Gene Dll4
Gene Accession No. Q9NR61
Gene Alias AOS6; delta 4; delta ligand 4; delta like canonical Notch ligand 4; Delta4; delta-like 4 (Drosophila); delta-like 4 homolog; delta-like 4 protein; delta-like protein 4; DLL4; Drosophila Delta homolog 4; hdelta2; MGC126344; notch ligand delta-2; notch ligand DLL4; UNQ1895/PRO4341
Gene Symbols Dll4
Host Species Rabbit
Immunogen Recombinant Human DLL4 protein (Ser 27-Pro524).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54567
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.