Learn More
DLX1 Monoclonal Antibody (OTI1A2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA811643
Encompass_Preferred
 View more versions of this product
Product Code. 17585774
  • € 540.87 / 100µL
Description
This gene encodes a member of a homeobox transcription factor gene family similiar to the Drosophila distal-less gene. The encoded protein is localized to the nucleus where it may function as a transcriptional regulator of signals from multiple TGF-{beta} superfamily members. The encoded protein may play a role in the control of craniofacial patterning and the differentiation and survival of inhibitory neurons in the forebrain. This gene is located in a tail-to-tail configuration with another member of the family on the long arm of chromosome 2. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications
Specifications
DLX1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P56177
DLX1
Human recombit protein fragment corresponding to amino acids 1-255 of human DLX1 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI1A2
Unconjugated
DLX1
distal-less homeobox 1
Mouse
Affinity Chromatography
RUO
1745
-20° C, Avoid Freeze/Thaw Cycles