missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLX1 Monoclonal Antibody (OTI3C5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811632S
This item is not returnable.
View return policy
Description
DLX1 Monoclonal antibody specifically detects DLX1 in Human samples. It is validated for Western BlotSpecifications
| DLX1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P56177 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1745 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI3C5 | |
| Unconjugated | |
| DLX1 | |
| DII B; distal-less homeo box 1; distal-less homeobox 1; Dlx; Dlx1; Dlx-1; homeobox Dlx1; homeobox protein DLX-1 | |
| Human recombinant protein fragment corresponding to amino acids 1-255 of human DLX1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction