missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLX3 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97552AF647
This item is not returnable.
View return policy
Description
DLX3 Polyclonal specifically detects DLX3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| DLX3 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| AI4, distal-less homeo box 3, distal-less homeobox 3, homeobox protein DLX-3, TDO | |
| Produced in rabbits immunized with E. coli-derived Human DLX3 fragment. | |
| 0.1 mL | |
| Growth and Development, Neuronal Cell Markers, Stem Cell Markers | |
| 1747 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction