missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dlx6 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97544C
This item is not returnable.
View return policy
Description
Dlx6 Polyclonal specifically detects Dlx6 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| Dlx6 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| distal-less homeo box 6, distal-less homeobox 6, homeobox protein DLX-6, MGC125282, MGC125283, MGC125284, MGC125285 | |
| Produced in rabbits immunized with E. coli-derived Human Dlx6 fragment. | |
| 0.1 mL | |
| Neuroscience | |
| 1750 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunofluorescence | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction