missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DLX6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566396
This item is not returnable.
View return policy
Description
Immunogen sequence: GNSYNHRSLAA YPYMSHSQHS PYLQSYHNSS AAAQTRGDDT DQQKTTVIEN GEIRF Highest antigen sequence identity to the following orthologs - mouse 100%, rat 75%.
The DLX6 gene encodes a member of a homeobox transcription factor gene family similiar to the Drosophila distal-less gene. This family is comprised of at least 6 different members that encode proteins with roles in forebrain and craniofacial development. This gene is in a tail-to-tail configuration with another member of the family on the long arm of chromosome 7.
Specifications
| DLX6 | |
| Polyclonal | |
| Unconjugated | |
| DLX6 | |
| distal-less homeo box 6; distal-less homeobox 6; distal-less homeobox 6-like; DLX6; Homeobox protein DLX-6; RGD1561539 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1750 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P56179 | |
| DLX6 | |
| Recombinant Human DLX6. Recombinant protein control fragment (Product #RP-107070). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction