missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DMAP1 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-256JF549
This item is not returnable.
View return policy
Description
DMAP1 Polyclonal antibody specifically detects DMAP1 in Human samples. It is validated for Western Blot
Specifications
| DMAP1 | |
| Polyclonal | |
| Western Blot | |
| DNA methyltransferase 1 associated protein 1, DNA methyltransferase 1-associated protein 1, DNMAP1DNMT1-associated protein 1, DNMT1 associated protein 1, DNMTAP1, EAF2, FLJ11543, KIAA1425DKFZp686L09142, MEAF2, SWC4 | |
| The immunogen recognized by this antibody maps to a region between residues 400 and 450 of human DNA Methyltransferase 1 Associated Protein 1 using the numbering given in entry NP_061973.1 (GeneID 55929). | |
| 0.1 mL | |
| Apoptosis, Chromatin Research, DNA Repair, MAP Kinase Signaling | |
| 55929 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction