missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DMAP1 Recombinant Rabbit Monoclonal Antibody (5Z8C8)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA537907
This item is not returnable.
View return policy
Description
Positive Samples: HeLa, PC-12, HepG2 Immunogen sequence: RSDLVLLYEL KQACANCEYE LQMLRHRHEA LARAGVLGGP ATPASGPGPA SAEPAVTEPG LGPDPKDTII DVVGAPLTPN SRKRRESASS SSSVKKAKKP.
DNA methyltransferase 1-associated protein 1 (DMAP1) is involved in the repression or activation of transcription. The encoded protein can independently repress transcription and is targeted to replication foci throughout S phase by interacting directly with the N-terminus of DNA methyltransferase 1. During late S phase, histone deacetylase 2 is added to this complex, providing a means to deacetylate histones in transcriptionally inactive heterochromatin following replication. The protein also specifically potentiates DAXX-mediated repression of glucocorticoid receptor-dependent transcription. The encoded protein is also a component of the nucleosome acetyltransferase of H4 complex and interacts with the transcriptional corepressor tumor susceptibility gene 101 and the pro-apoptotic death-associated protein 6, among others. DMAP1 is found in a wide variety of living organisms from yeasts to humans. The human DMAP1 gene is localized on chromosome 1p34.
Specifications
| DMAP1 | |
| Recombinant Monoclonal | |
| 2 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9JI44, Q9NPF5 | |
| DMAP1 | |
| A synthetic peptide corresponding to a sequence within amino acids 368-467 of human DMAP1 (Q9NPF5). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 5Z8C8 | |
| Unconjugated | |
| DMAP1 | |
| 1500016M21Rik; DKFZp686L09142; Dmap1; Dmtap1; DNA methyltransferase 1 associated protein 1; DNA methyltransferase 1-associated protein 1; DNMAP1; DNMT1 associated protein 1; DNMT1 associated protein-1; DNMT1-associated protein 1; DNMTAP1; EAF2; FLJ11543; KIAA1425; MAT1-mediated transcriptional repressor; MEAF2; mKIAA1425; Mmtr; RP5-891H21.2; SWC4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 298447, 55929, 66233 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction