missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DMC1 Antibody (2H12/4), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2617F
This item is not returnable.
View return policy
Description
DMC1 Monoclonal antibody specifically detects DMC1 in Human, Mouse, Rat, Bovine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence
Specifications
| DMC1 | |
| Monoclonal | |
| FITC | |
| disrupted meiotic cDNA1, yeast, homolog of, DMC1 dosage suppressor of mck1 homolog, meiosis-specific homologousrecombination (yeast), DMC1 homologue, HsLim15, LIM15yeast homolog) meiosis-specific homologousrecombination, meiotic recombination protein DMC1/LIM15 homolog, MGC150472, MGC150473 | |
| Recombinant fusion protein (Human). | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Cycle and Replication, Chromatin Research, DNA Repair, Homologous Recombination, Tumor Suppressors | |
| 11144 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
| 2H12/4 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction