missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DMC1 Antibody (2H12/4), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2617H
This item is not returnable.
View return policy
Description
DMC1 Monoclonal antibody specifically detects DMC1 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| DMC1 | |
| Monoclonal | |
| HRP | |
| disrupted meiotic cDNA1, yeast, homolog of, DMC1 dosage suppressor of mck1 homolog, meiosis-specific homologousrecombination (yeast), DMC1 homologue, HsLim15, LIM15yeast homolog) meiosis-specific homologousrecombination, meiotic recombination protein DMC1/LIM15 homolog, MGC150472, MGC150473 | |
| Recombinant fusion protein (Human). | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Cycle and Replication, Chromatin Research, DNA Repair, Homologous Recombination, Tumor Suppressors | |
| 11144 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| 2H12/4 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction