Learn More
DMC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13714-1-AP-150UL
 View more versions of this product
Product Code. 16812074
  • € 410.00 / 150µL
Description
DMC1 is essential for meiotic homologous recombination. Genetic recombination in meiosis plays an important role in generating diversity of genetic information. The product of this protein is structurally and evolutionary related to the products of the yeast RAD51 and E. coli RecA genes.
Specifications
Specifications
DMC1
Polyclonal
Unconjugated
DMC1
DMC1, DMC1H, HsLim15, LIM15
Rabbit
Antigen Affinity Chromatography
RUO
11144, 13404, 362960
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q14565, Q61880
Dmc1
DMC1 Fusion Protein Ag4653
150 μL
Primary
Human, Mouse, Rat