missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DMGDH Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113450
This item is not returnable.
View return policy
Description
DMGDH Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
This gene encodes an enzyme involved in the catabolism of choline, catalyzing the oxidative demethylation of dimethylglycine to form sarcosine. The enzyme is found as a monomer in the mitochondrial matrix, and uses flavin adenine dinucleotide and folate as cofactors. Mutation in this gene causes dimethylglycine dehydrogenase deficiency, characterized by a fishlike body odor, chronic muscle fatigue, and elevated levels of the muscle form of creatine kinase in serum. Alternative splicing results in multiple transcript variants.
Specifications
| DMGDH | |
| Polyclonal | |
| Unconjugated | |
| DMGDH | |
| 1200014D15Rik; AI787269; dimethylglycine dehydrogenase; dimethylglycine dehydrogenase precursor; dimethylglycine dehydrogenase, mitochondrial; Dmgdh; DMGDHD; EC 1.5.99.2; M2GD; ME2GLYDH | |
| Rabbit | |
| Protein G | |
| RUO | |
| 29958, 74129 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 3.34 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9DBT9, Q9UI17 | |
| DMGDH | |
| Recombinant Human Dimethylglycine dehydrogenase, mitochondrial protein (429-524AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction