Learn More
DMKN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16805834 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16805834 150 μL 150µL
16815834 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16805834 Supplier Proteintech Supplier No. 162521AP150UL
Rabbit Polyclonal Antibody
The function of this protein is still unknown. The human ZD52F10 gene is up-regulated in colon cancer metastasis. It encode the protein (dermokine), which is a new stratified epithelium-secreted protein.
Specifications
Antigen DMKN
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DMKN
Gene Accession No. Q6E0U4, Q6P253
Gene Alias dermokine, DMKN, UNQ729, ZD52F10
Gene Symbols Dmkn
Host Species Rabbit
Immunogen DMKN Fusion Protein Ag9242
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361548, 73712, 93099
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG