missing translation for 'onlineSavingsMsg'
Learn More

DMKN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16815834 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16815834 20 μL 20µL
16805834 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16815834 Supplier Proteintech Supplier No. 162521AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The function of this protein is still unknown. The human ZD52F10 gene is up-regulated in colon cancer metastasis. It encode the protein (dermokine), which is a new stratified epithelium-secreted protein.

Specifications

Antigen DMKN
Applications Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DMKN
Gene Accession No. Q6E0U4, Q6P253
Gene Alias dermokine, DMKN, UNQ729, ZD52F10
Gene Symbols Dmkn
Host Species Rabbit
Immunogen DMKN Fusion Protein Ag9242
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361548, 73712, 93099
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.