missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DMP-1 Antibody (001), DyLight 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90020UV
This item is not returnable.
View return policy
Description
DMP-1 Monoclonal specifically detects DMP-1 in Human samples. It is validated for ELISA.
Specifications
| DMP-1 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 1758 | |
| Human | |
| Purified |
| ELISA | |
| 1 | |
| ELISA | |
| ARHP, ARHR, dentin matrix acidic phosphoprotein, dentin matrix acidic phosphoprotein 1, Dentin matrix protein 1, DMP-1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human DMP-1 (Q13316-1; Met1-Tyr513). | |
| 0.1 mL | |
| Primary | |
| No cross-reactivity in ELISA with: Human CD44 Human CFH | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction