missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DMPO Nitrone Adduct Antibody (N1664A), Novus Biologicals™
Mouse Monoclonal Antibody
€ 174.00 - € 500.00
Specifications
| Antigen | DMPO Nitrone Adduct |
|---|---|
| Clone | N1664A |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18242425
|
Novus Biologicals
NBP2-59306 |
100 μg |
€ 500.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18607366
|
Novus Biologicals
NBP2-59306-25ug |
25 μg |
€ 174.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DMPO Nitrone Adduct Monoclonal specifically detects DMPO Nitrone Adduct in Mouse, Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Microarray.Specifications
| DMPO Nitrone Adduct | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
| Unconjugated | |
| Mouse | |
| 2,2-dimethyl-1-oxido-3,4-dihydropyrrol-1-ium, 2,2-Dimethyl-3,4-dihydro-2H-pyrrole 1-oxide, 5,5-Dimethyl-1-pyrroline N-oxide, 5,5-Dimethyl-1-pyrroline-1-oxide | |
| IgG1 | |
| Protein G purified |
| N1664A | |
| Monoclonal | |
| Purified | |
| RUO | |
| 5,5-dimethyl-2-(8-octanoic acid)-1-pyrrolone-N-oxide conjugated to Ovalbumin | |
| Primary | |
| Recognizes DMPO, DMPO-octanoic acid, DMPO-protein adducts and DMPO-DNA adducts. Does not cross react with non-adducted proteins or DNA. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title