missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DMRT1 Antibody (OTI1G10), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70582APC
This item is not returnable.
View return policy
Description
DMRT1 Monoclonal antibody specifically detects DMRT1 in Human samples. It is validated for Western Blot
Specifications
| DMRT1 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 1761 | |
| Store at 4C in the dark. | |
| IgG2b |
| Western Blot | |
| OTI1G10 | |
| Western Blot | |
| DM domain expressed in testis 1, DMT1DM domain expressed in testis protein 1, doublesex and mab-3 related transcription factor 1, doublesex- and mab-3-related transcription factor 1 | |
| Human recombinant protein fragment corresponding to amino acids 1-210 of human DMRT1(NP_068770) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction