missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DMRT1 Monoclonal Antibody (OTI2A10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807523S
This item is not returnable.
View return policy
Description
DMRT1 Monoclonal antibody specifically detects DMRT1 in Human samples. It is validated for Western BlotSpecifications
| DMRT1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y5R6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1761 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI2A10 | |
| Unconjugated | |
| DMRT1 | |
| CT154; DM domain expressed in testis 1; DM domain expressed in testis protein 1; Dmrt1; DMT1; Doublesex and mab-3 related transcription factor 1; doublesex- and mab-3-related transcription factor 1; Unknown (protein for MGC:134119) | |
| Human recombinant protein fragment corresponding to amino acids 1-210 of human DMRT1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction