missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DMRT1 Monoclonal Antibody (OTI2H1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807746
This item is not returnable.
View return policy
Description
This gene is found in a cluster with two other members of the gene family, having in common a zinc finger-like DNA-binding motif (DM domain). The DM domain is an ancient, conserved component of the vertebrate sex-determining pathway that is also a key regulator of male development in flies and nematodes. This gene exhibits a gonad-specific and sexually dimorphic expression pattern. Defective testicular development and XY feminization occur when this gene is hemizygous.Specifications
| DMRT1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y5R6 | |
| DMRT1 | |
| Human recombit protein fragment corresponding to amino acids 1-210 of human DMRT1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2H1 | |
| Unconjugated | |
| DMRT1 | |
| CT154, DMT1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1761 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction