missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DMRT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579160
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat testis tissue, mouse testis tissue. IHC: mouse testis tissue, rat testis tissue, human testis tissue.
This gene is found in a cluster with two other members of the gene family, having in common a zinc finger-like DNA-binding motif (DM domain). The DM domain is an ancient, conserved component of the vertebrate sex-determining pathway that is also a key regulator of male development in flies and nematodes. This gene exhibits a gonad-specific and sexually dimorphic expression pattern. Defective testicular development and XY feminization occur when this gene is hemizygous.
Specifications
| DMRT1 | |
| Polyclonal | |
| Unconjugated | |
| DMRT1 | |
| CT154; DM domain expressed in testis 1; DM domain expressed in testis protein 1; Dmrt1; DMT1; Doublesex and mab-3 related transcription factor 1; doublesex- and mab-3-related transcription factor 1; Unknown (protein for MGC:134119) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114498, 1761, 50796 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| Q9QZ59, Q9Y5R6 | |
| DMRT1 | |
| A synthetic peptide corresponding to a sequence at the N-terminus of human DMRT1 (98-128aa RDCQCKKCNLIAERQRVMAAQVALRRQQAQE). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction