missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DMRT2 Polyclonal Antibody

Product Code. 15847303
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15847303 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15847303 Supplier Invitrogen™ Supplier No. PA568891

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 79%; Dog: 86%; Guinea Pig: 86%; Human: 86%; Mouse: 100%; Rabbit: 86%; Rat: 86%.

Gonadal dysgenesis in 9p-deleted individuals might be due to combined hemizygosity of DMRT1 and DMRT2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DMRT2
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DMRT2
Gene Accession No. Q8BG36
Gene Alias dmrt2; doublesex and mab-3 related transcription factor 2; doublesex- and mab-3-related transcription factor 2; Doublesex-like 2 protein; doublesex-like 2 protein (DSXL-2); DSXL2; DSXL-2; LOW QUALITY PROTEIN: doublesex- and mab-3-related transcription factor 2; Terra; terra homolog; terra like protein antibody; terra-like protein
Gene Symbols DMRT2
Host Species Rabbit
Immunogen synthetic peptide directed towards the C terminal region of mouse DMRT2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 226049
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.