missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DMRT3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585040
This item is not returnable.
View return policy
Description
Immunogen sequence: MNGYGSPYLY MGGPVSQPPR APLQRTPKC Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.
Probable transcription factor that plays a role in configuring the spinal circuits controlling stride in vertebrates. Involved in neuronal specification within specific subdivision of spinal cord neurons and in the development of a coordinated locomotor network controlling limb movements. May regulate transcription during sexual development. [UniProt].
Specifications
| DMRT3 | |
| Polyclonal | |
| Unconjugated | |
| DMRT3 | |
| DMRT3; DMRTA3; DMRT-like family A3; Doublesex and mab-3 related transcription factor 3; doublesex- and mab-3-related transcription factor 3; testis-specific protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 58524 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NQL9 | |
| DMRT3 | |
| Recombinant protein corresponding to Human DMRT3. Recombinant protein control fragment (Product #RP-108690). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction