missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DMRTB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100784
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total DMRTB1.
DMRTB1 gene ontology annotations related to this gene include DNA-binding transcription factor activity.
Specifications
| DMRTB1 | |
| Polyclonal | |
| Unconjugated | |
| DMRTB1 | |
| DMRT like family B with proline rich C-terminal 1; Dmrt6; Dmrtb1; DMRT-like family B with proline-rich C-terminal 1; DMRT-like family B with proline-rich C-terminal, 1; doublesex and mab-3 related transcription factor 6; doublesex- and mab-3-related transcription factor 6; Doublesex- and mab-3-related transcription factor B1; proline-rich protein 13; Prp13; RGD1560921 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 56296, 63948 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A2A9I7, Q96MA1 | |
| DMRTB1 | |
| A synthesized peptide derived from human DMRTB1(Accession Q96MA1), corresponding to amino acid residues F294-D342. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction