missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB3A Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68052-1-IG-150UL
This item is not returnable.
View return policy
Description
The RAB3A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RAB3A. This antibody recognizes human, mouse, rat, pig, rabbit, chicken antigen. The RAB3A antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| RAB3A | |
| Monoclonal | |
| 500 μg/mL | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 5864 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG2b |
| Western Blot, Immunohistochemistry, ELISA | |
| 1B12E9 | |
| Unconjugated | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat, Pig, Rabbit, Chicken | |
| 25 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction