missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DMTF1 Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova PAB30039.100uL
This item is not returnable.
View return policy
Description
Rabbit polyclonal antibody raised against synthetic peptide of human DMTF1.
- High specificity
- High reactivity
Specifications
| DMTF1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of human DMTF1. | |
| In PBS (2% sucrose, 0.09% sodium azide). | |
| DMP1/DMTF/FLJ25188/FLJ41265/FLJ76054/hDMP1 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 9988 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-8 ug/mL) Western Blot (0.2-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| NM_001142326 | |
| DMTF1 | |
| A synthetic peptide corresponding to C-terminus of human DMTF1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction