missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Antibody (DSD/958), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11590PCP
This item is not returnable.
View return policy
Description
DNA Monoclonal antibody specifically detects DNA in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| DNA | |
| Monoclonal | |
| PerCP | |
| Double stranded deoxyribonucleic acid, Double stranded DNA, dsDNA | |
| Nuclei of Burkitt's cells | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG3 κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DSD/958 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction