missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNA Ligase I Recombinant Rabbit Monoclonal Antibody (K01_2N08)

Product Code. 30825811
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30825811 50 μL
30830046 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30825811 Supplier Invitrogen™ Supplier No. MA565568

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

DNA Ligase I Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Western Blot

The protein encoded by this gene is involved in the synthesis of asparagine. This gene complements a mutation in the temperature-sensitive hamster mutant ts11, which blocks progression through the G1 phase of the cell cycle at nonpermissive temperature. There are three alternatively spliced transcript variants encoding the same protein described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA Ligase I
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_2N08
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene LIG1
Gene Accession No. P18858, Q9JHY8
Gene Alias AL033288; DNA ligase (ATP) 1; DNA ligase 1; DNA ligase I; LIG1; Lig-1; ligase 1; ligase I, DNA, ATP-dependent; LigI; MGC117397; MGC130025; polydeoxyribonucleotide synthase [ATP] 1
Gene Symbols LIG1
Host Species Rabbit
Immunogen A synthetic peptide of human LIG1.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3978, 81513
Target Species Human, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.