missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNA Ligase III Monoclonal Antibody (1F3)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA123191

Product Code. 11511460

  • € 588.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: 293T, A431, HeLa, HepG2. The predicted molecular weight of MA1-23191 is 100 kDa.

There are four distinct ligase proteins found in mammalian cells. The activity of these DNA ligases is required for the joining of DNA strand breaks that occur during DNA replication, repair and recombination. DNA Ligase III gene product migrates as a 100 kDa (a-form) protein in somatic tissues, is ubiquitously expressed and complexes with XRCC-1 and a 97 kDa (beta form) protein that is found in testis. DNA Ligase III is involved in rejoining DNA single-strand breaks and in the base excision repair pathway.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DNA Ligase III
Monoclonal
1 mg/mL
PBS with no preservative
P49916, P97386
LIG3
Recombinant full-length human DNA Ligase III-b.
100 μL
Primary
Human, Mouse, Chicken
Antibody
IgG1
Immunoprecipitation, Neutralization, Western Blot, Proximity Ligation Assay, Immunocytochemistry
1F3
Unconjugated
LIG3
D11Wsu78e; DNA ligase (ATP) 3; DNA ligase 3; DNA ligase III; hypothetical protein; LIG2; LIG3; ligase II, DNA, ATP-dependent; ligase III, DNA, ATP-dependent; polydeoxyribonucleotide synthase [ATP] 3; Unknown (protein for MGC:134139)
Mouse
Protein G
RUO
16882, 3980, 417530
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.